Invitrogen
Manufacturer Code:PA559027
Catalog # PIPA559027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DNA Primase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA primase 1; DNA primase 1 DNA primase 49 kDa subunit DNA primase small subunit EC 2.7.7.- PRI1; DNA primase 49 kDa subunit; DNA primase small subunit; DNA primase subunit 48; p49; primase p49 subunit; primase polypeptide 1, 49kDa; primase, DNA, polypeptide 1 (49kDa)
Gene Aliases: p49; PRIM1
UniProt ID: (Human) P49642
Entrez Gene ID: (Human) 5557
Molecular Function:
RNA binding protein
nucleic acid binding
primase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.