Invitrogen
Manufacturer Code:PA539885
Catalog # PIPA539885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human DNA Polymerase theta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C |
DNA Polymerase theta Antibody, PA5-39885, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).
Protein Aliases: DNA polymerase eta; DNA polymerase theta; POLH; polymerase (DNA directed), theta; polymerase (DNA) theta
Gene Aliases: POLH; POLQ; PRO0327
UniProt ID: (Human) O75417
Entrez Gene ID: (Human) 10721
Molecular Function: DNA binding protein DNA-directed DNA polymerase exodeoxyribonuclease hydrolase nuclease nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.