Novus Biologicals
Manufacturer Code:NB100476AF750
Catalog # NB10476A750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 607 |
Immunogen | Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 3'-5' exodeoxyribonuclease; CDC2 CDC2 homolog DNA polymerase delta catalytic subunit DNA polymerase subunit delta p125 DNA-directed polymerase delta 1 EC 2.7.7.7 POLD polymerase (DNA directed) delta 1 catalytic subunit (125kD) polymerase (DNA directed) delta 1 catalytic subunit 125kDa; CDC2 homolog; DNA polymerase delta catalytic subunit; DNA polymerase subunit delta p125; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit
Gene Aliases: CDC2; CRCS10; MDPL; POLD; POLD1
UniProt ID: (Human) P28340
Entrez Gene ID: (Human) 5424
Molecular Function: exodeoxyribonuclease nuclease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.