Invitrogen
Manufacturer Code:MA124221
Catalog # PIMA124221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6196 |
Immunogen | Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3'-5' exodeoxyribonuclease; CDC2 homolog; CDC2 POLD; DNA polymerase delta catalytic subunit; DNA polymerase subunit delta p125; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit
Gene Aliases: CDC2; CRCS10; MDPL; POLD; POLD1
UniProt ID: (Human) P28340
Entrez Gene ID: (Human) 5424
Molecular Function: exodeoxyribonuclease nuclease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.