Invitrogen
Manufacturer Code:MA513235
Catalog # PIMA513235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 18-2 |
Immunogen | Human DNA-dependent Protein Kinase purified from HeLa cells |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-dependent protein kinase catalytic subunit; DNA-PK catalytic subunit; DNA-PK catalytic subunit DNA-dependent protein kinase catalytic subunit hyper-radiosensitivity of murine scid mutation complementing 1 p460; DNPK1; hyper-radiosensitivity of murine scid mutation, complementing 1; p460
Gene Aliases: DNA-PKcs; DNAPK; DNPK1; HYRC; HYRC1; IMD26; p350; PRKDC; XRCC7
UniProt ID: (Human) P78527
Entrez Gene ID: (Human) 5591, (Rat) 360748
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.