Invitrogen
Manufacturer Code:PA123197
Catalog # PIPA123197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human DNA PKcs |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-dependent protein kinase catalytic subunit; DNA-PK catalytic subunit; DNPK1; doxorubicin nephropathy; hyper-radiosensitivity of murine scid mutation, complementing 1; p460; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; severe combined immunodeficiency
Gene Aliases: AI326420; AU019811; DNA-PKcs; DNAPDcs; DNAPK; DNPK1; DOXNPH; dxnph; HYRC; HYRC1; I79_017613; IMD26; p350; p460; PRKDC; scid; slip; XRCC7
UniProt ID: (Human) P78527, (Mouse) P97313
Entrez Gene ID: (Human) 5591, (Chinese hamster) 100770748, (Mouse) 19090, (Rat) 360748
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.