Invitrogen
Manufacturer Code:MA535109
Catalog # PIMA535109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0595 |
Immunogen | A synthesized peptide derived from human DNA Ligase IV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 5830471N16Rik BOS_12728 DNA ligase 4 DNA ligase IV LIG4 LIG4S Polydeoxyribonucleotide synthase [ATP] 4 tiny; DNA joinase; DNA ligase 4; DNA ligase IV; DNA repair enzyme; ligase IV, DNA, ATP-dependent; Polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase
Gene Aliases: 5830471N16Rik; LIG4; LIG4S; tiny
UniProt ID: (Human) P49917, (Mouse) Q8BTF7
Entrez Gene ID: (Human) 3981, (Mouse) 319583
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.