Invitrogen
Manufacturer Code:PA570056
Catalog # PIPA570056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human DNA Ligase III |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; LIG2 DNA ligase 3 ligase II DNA ATP-dependent polydeoxyribonucleotide synthase &lsqbATP] 3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; Polydeoxyribonucleotide synthase [ATP] 3
Gene Aliases: D11Wsu78e; LIG2; LIG3
UniProt ID: (Human) P49916
Entrez Gene ID: (Rabbit) 100344941, (Pig) 100626381, (Horse) 100071671, (Human) 3980, (Dog) 491145, (Guinea pig) 100716163, (Rat) 303369, (Mouse) 16882
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.