Novus Biologicals
Manufacturer Code:NBP29271920UL
Catalog # NB027308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 730-1009 of human LIG3 (NP_039269.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA ligase 3; DNA ligase 3 DNA ligase III EC 6.5.1.1 LIG2 ligase II DNA ATP-dependent ligase III DNA ATP-dependent Polydeoxyribonucleotide synthase [ATP] 3; DNA ligase III; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; Polydeoxyribonucleotide synthase [ATP] 3
Gene Aliases: LIG2; LIG3
UniProt ID: (Human) P49916
Entrez Gene ID: (Human) 3980
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.