Invitrogen
Manufacturer Code:PA599788
Catalog # PIPA599788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DMRTC2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933432E21Rik DMRT like family C2 Dmrt7 DMRTC2 DMRT-like family C2 doublesex and mab-3 related transcription factor 7 doublesex and mab-3 related transcription factor like family C2 Doublesex- and mab-3-related transcription factor 7 Doublesex- and mab-3-related transcription factor C2; DMRT-like family C2; Doublesex- and mab-3-related transcription factor C2
Gene Aliases: DMRTC2
UniProt ID: (Human) Q8IXT2
Entrez Gene ID: (Human) 63946
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.