Invitrogen
Manufacturer Code:PA575746
Catalog # PIPA575746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human DMRTC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMRT-like family C2; doublesex and mab-3 related transcription factor 7; Doublesex- and mab-3-related transcription factor 7; Doublesex- and mab-3-related transcription factor C2; Doublesex- and mab-3-related transcription factor C2 DMRTC2
Gene Aliases: 4933432E21Rik; Dmrt7; DMRTC2
UniProt ID: (Human) Q8IXT2, (Mouse) Q8CGW9
Entrez Gene ID: (Human) 63946, (Mouse) 71241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.