Invitrogen
Manufacturer Code:PA568863
Catalog # PIPA568863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse DMRTC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMRT-like family C2; doublesex and mab-3 related transcription factor 7; doublesex and mab-3 related transcription factor like family C2; Doublesex- and mab-3-related transcription factor 7; Doublesex- and mab-3-related transcription factor C2
Gene Aliases: 4933432E21Rik; Dmrt7; DMRTC2
UniProt ID: (Human) Q8IXT2, (Bovine) Q32LE6, (Mouse) Q8CGW9
Entrez Gene ID: (Rabbit) 100340563, (Horse) 100069866, (Human) 63946, (Bovine) 539134, (Dog) 102155486, (Mouse) 71241, (Guinea pig) 100725551, (Rat) 499100
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.