Novus Biologicals
Manufacturer Code:NBP180248
Catalog # NBP180248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human DMRTC2. Peptide sequence MDPSETAALHHCSADSSPADEARVPQSTELIPRRPVSRSPTCARCRNHGV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMRT-like family C2; DMRT-like family C2 doublesex- and mab-3-related transcription factor C2; Doublesex- and mab-3-related transcription factor C2
Gene Aliases: DMRTC2
UniProt ID: (Human) Q8IXT2
Entrez Gene ID: (Human) 63946
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.