Invitrogen
Manufacturer Code:PA535234
Catalog # PIPA535234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 487-515 amino acids from the C-terminal region of human DMRTA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMRT-like family A2; DMRT5; DMRTA2 DMRT5 Doublesex- and mab-3-related transcription factor A2 Doublesex- and mab-3-related transcription factor 5; doublesex and mab-3 related transcription factor like family A2; Doublesex- and mab-3-related transcription factor 5; Doublesex- and mab-3-related transcription factor A2
Gene Aliases: DMRT5; DMRTA2
UniProt ID: (Human) Q96SC8, (Mouse) A2A9A2
Entrez Gene ID: (Human) 63950, (Mouse) 242620, (Zebrafish) 474350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.