Novus Biologicals
Manufacturer Code:NBP2828630.1ML
Catalog # NB030792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human DMRT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: doublesex and mab-3 related transcription factor 2 doublesex- and mab-3-related transcription factor 2 Doublesex-like 2 protein DSXL2 DSXL-2 terra-like protein; Doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; DSXL-2; terra-like protein
Gene Aliases: DMRT2; DSXL2
UniProt ID: (Human) Q9Y5R5
Entrez Gene ID: (Human) 10655
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.