Invitrogen
Manufacturer Code:PA599271
Catalog # PIPA599271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DMPK corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DM DM protein kinase DM1 DM1 protein kinase Dm15 DM1PK DMK DM-kinase DMPK dystrophia myotonica kinase B15 dystrophia myotonica protein kinase dystrophia myotonica-protein kinase Mdpk MT-PK myotonic dystrophy associated protein kinase myotonic dystrophy protein kinase myotonin protein kinase A myotonin-protein kinase thymopoietin homolog; DM protein kinase; DM-kinase; DM1 protein kinase; DMK; DMPK; dystrophia myotonica-protein kinase; MT-PK; myotonic dystrophy associated protein kinase; Myotonic dystrophy protein kinase; myotonin protein kinase A; Myotonin-protein kinase; thymopoietin homolog
Gene Aliases: DM; DM1; DM1PK; DMK; DMPK; MDPK; MT-PK
UniProt ID: (Human) Q09013
Entrez Gene ID: (Human) 1760
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.