Invitrogen
Manufacturer Code:PA542121
Catalog # PIPA542121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DMPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DM protein kinase; DM-kinase; DM1 protein kinase; DMK; DMPK; dystrophia myotonica kinase, B15; dystrophia myotonica protein kinase; dystrophia myotonica-protein kinase; Dystrophia myotonica-protein kinase DM DM1 DM1PK DMK MDPK MT-PK; MDPK; MT-PK; myotonic dystrophy associated protein kinase; Myotonic dystrophy protein kinase; myotonin protein kinase A; Myotonin-protein kinase; thymopoietin homolog
Gene Aliases: DM; DM1; Dm15; DM1PK; DMK; DMPK; MDPK; MT-PK
UniProt ID: (Human) Q09013, (Mouse) P54265
Entrez Gene ID: (Dog) 484436, (Horse) 100146609, (Human) 1760, (Rat) 308405, (Guinea pig) 100713533, (Mouse) 13400
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.