Proteintech
Manufacturer Code:248131AP150UL
Catalog # 50-173-4558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DMGDH Fusion Protein Ag20529 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dimethylglycine dehydrogenase DMGDH DMGDHD M2GD ME2GLYDH; Dimethylglycine dehydrogenase, mitochondrial; ME2GLYDH
Gene Aliases: 1200014D15Rik; AI787269; DMGDH; DMGDHD; ME2GLYDH
UniProt ID: (Human) Q9UI17, (Mouse) B1B1D0
Entrez Gene ID: (Human) 29958, (Mouse) 74129, (Rat) 245961
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.