Novus Biologicals
Manufacturer Code:NB1002687
Catalog # NB1002687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B10/2 |
Immunogen | Recombinant fusion DMC1 protein (expressed as 6xHis fusion in E. coli and band cut out of gel of insoluble pellet). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: disrupted meiotic cDNA1 yeast homolog of DMC1 dosage suppressor of mck1 homolog meiosis-specific homologousrecombination (yeast) DMC1 homologue HsLim15 LIM15yeast homolog) meiosis-specific homologousrecombination meiotic recombination protein DMC1/LIM15 homolog MGC150472 MGC150473; disrupted meiotic cDNA1, yeast, homolog of; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; Meiotic recombination protein DMC1/LIM15 homolog
Gene Aliases: dJ199H16.1; DMC1; DMC1H; LIM15
UniProt ID: (Human) B4DMW6
Entrez Gene ID: (Human) 11144
Molecular Function: DNA binding protein DNA strand-pairing protein hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.