Proteintech
Manufacturer Code:137141AP150UL
Catalog # 50-172-8953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DMC1 Fusion Protein Ag4653 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: disrupted meiotic cDNA 1 homolog; disrupted meiotic cDNA1, yeast, homolog of; DMC1 DMC1H HsLim15 LIM15; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; Meiotic recombination protein DMC1/LIM15 homolog
Gene Aliases: dJ199H16.1; DMC1; DMC1H; LIM15; Mei11; sgdp
UniProt ID: (Human) B4DMW6, (Mouse) Q61880
Entrez Gene ID: (Human) 11144, (Mouse) 13404, (Rat) 362960
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.