Invitrogen
Manufacturer Code:PA572622
Catalog # PIPA572622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 167-194 amino acids from the C-terminal region of DANRE dlx6a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: distal-less homeobox gene 6a; Distal-less homeobox protein 6a; DLX-6; Homeobox protein Dlx6a; Homeobox protein Dlx6a Distal-less homeobox protein 6a DLX-6 dlx6a dlx6
Gene Aliases: dlx6; dlx6a
UniProt ID: (Zebrafish) Q98877
Entrez Gene ID: (Zebrafish) 30586
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.