Invitrogen
Manufacturer Code:PA575312
Catalog # PIPA575312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 84-104 of Human DLX4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta protein 1; DII D; distal-less homeo box 7; distal-less homeo box 9; Homeobox protein DLX-4; Homeobox protein DLX-4 Beta protein 1 Homeobox protein DLX-7 Homeobox protein DLX-8 DLX4 BP1 DLX7 DLX8 DLX9; Homeobox protein DLX-7; Homeobox protein DLX-8
Gene Aliases: BP1; Dlx-4; DLX4; DLX7; DLX8; DLX9; OFC15
UniProt ID: (Human) Q92988, (Mouse) P70436
Entrez Gene ID: (Human) 1748, (Rat) 303469, (Mouse) 13394
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.