Invitrogen
Manufacturer Code:PA540506
Catalog # PIPA540506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DLX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DII C; Distal-less homeobox 3 AI4 TDO; Homeobox protein DLX-3
Gene Aliases: AI4; AV237891; Dlx-3; DLX3; TDO
UniProt ID: (Human) O60479, (Mouse) Q64205
Entrez Gene ID: (Sheep) 101109372, (Human) 1747, (Bovine) 784366, (Dog) 491073, (Rabbit) 100342325, (Horse) 100146218, (Guinea pig) 100735447, (Mouse) 13393, (Rat) 287638
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.