Invitrogen
Manufacturer Code:PA523398
Catalog # PIPA523398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (238 SASPSYLDDPTNSWYH 253) of DLX3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI4 TDO homeobox protein DLX-3; DII C; Homeobox protein DLX-3
Gene Aliases: AI4; AV237891; Dlx-3; DLX3; TDO
UniProt ID: (Human) O60479, (Mouse) Q64205
Entrez Gene ID: (Human) 1747, (Bovine) 784366, (Dog) 491073, (Horse) 100146218, (Mouse) 13393
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.