Invitrogen
Manufacturer Code:PA546974
Catalog # PIPA546974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse DLL4 Ser28-Pro525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Delta-like protein 4; Delta4; Drosophila Delta homolog 4; hdelta2 delta 4 delta ligand 4 delta-like 4 homolog delta-like 4 protein delta-like protein 4 delta4 drosophila Delta homolog 4 notch ligand DLL4 notch ligand delta-2
Gene Aliases: Delta4; Dll4
UniProt ID: (Mouse) Q9JI71
Entrez Gene ID: (Mouse) 54485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.