R&D Systems
Manufacturer Code:AF1389SP
Catalog # AF1389SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse DLL4 Ser28-Pro525 Accession # BAB18580 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: delta 4; delta 4 Delta 4 precursor delta ligand 4 delta4 delta-like 4 (Drosophila) delta-like 4 homolog delta-like 4 homolog (Drosophila) delta-like 4 protein delta-like protein 4 Drosophila Delta homolog 4 hdelta2 MGC126344 notch ligand delta-2 notch ligand DLL4; delta ligand 4; delta-like 4 homolog; delta-like 4 protein; Delta-like protein 4; Delta4; Drosophila Delta homolog 4; notch ligand delta-2; notch ligand DLL4
Gene Aliases: AOS6; DLL4; hdelta2; UNQ1895/PRO4341
UniProt ID: (Human) Q9NR61
Entrez Gene ID: (Human) 54567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.