Proteintech
Manufacturer Code:215841AP150UL
Catalog # 50-173-3500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DLL4 Fusion Protein Ag16140 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: delta 4; delta ligand 4; delta like 4 (Drosophila) Delta like protein 4 Delta4 DLL4 Drosophila Delta homolog 4 hdelta2; delta-like 4 homolog; delta-like 4 protein; Delta-like protein 4; Delta4; Drosophila Delta homolog 4; notch ligand delta-2; notch ligand DLL4
Gene Aliases: AOS6; Delta4; DLL4; hdelta2; UNQ1895/PRO4341
UniProt ID: (Human) Q9NR61, (Mouse) Q9JI71
Entrez Gene ID: (Human) 54567, (Mouse) 54485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.