Invitrogen
Manufacturer Code:PA523448
Catalog # PIPA523448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (124 IETWREELGDQIGGP 138) of DLL3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: delta-like 3; Delta-like protein 3; Delta3; Drosophila Delta homolog 3; M-Delta-3; SCDO1 delta-like protein 3 delta3 drosophila Delta homolog 3
Gene Aliases: DLL3; pu; pudgy; SCDO1
UniProt ID: (Human) Q9NYJ7, (Mouse) O88516
Entrez Gene ID: (Human) 10683, (Mouse) 13389
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.