Invitrogen
Manufacturer Code:PA5113841
Catalog # PIPA5113841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Human DLL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: delta like canonical Notch ligand 3 Delta3 delta-like 3 delta-like 3 (Drosophila) Delta-like protein 3 DLL3 Drosophila Delta homolog 3 M-Delta-3 pu pudgy SCDO1; delta-like 3; Delta-like protein 3; Delta3; Drosophila Delta homolog 3
Gene Aliases: DLL3; SCDO1
UniProt ID: (Human) Q9NYJ7
Entrez Gene ID: (Human) 10683
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.