Novus Biologicals
Manufacturer Code:NBP2122660.025MG
Catalog # NB019135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0247- 2X38 |
Immunogen | Human DLL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Delta delta (Drosophila)-like 1 Delta1 delta-like 1 (Drosophila) delta-like protein 1 DL1 Drosophila Delta homolog 1 H-Delta-1; Delta-like protein 1; Delta1; Drosophila Delta homolog 1; H-Delta-1
Gene Aliases: Delta; DELTA1; DL1; DLL1; UNQ146/PRO172
UniProt ID: (Human) O00548
Entrez Gene ID: (Human) 28514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.