Invitrogen
Manufacturer Code:MA530337
Catalog # PIMA530337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 306 |
Immunogen | Recombit Human DLL1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Delta-like protein 1; Delta1; DLL1 delta-like 1 (Drosophila) DELTA1 DL1 Delta OTTHUMP00000017690 delta (Drosophila)-like 1 delta-like 1 delta-like 1 protein; Drosophila Delta homolog 1; H-Delta-1
Gene Aliases: Delta; DELTA1; DL1; DLL1; UNQ146/PRO172
UniProt ID: (Human) O00548
Entrez Gene ID: (Human) 28514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.