R&D Systems
Manufacturer Code:MAB18181SP
Catalog # MAB18181SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 251123 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human DLL1 Ser22-Gly540 Accession # AAG09716 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Delta delta (Drosophila)-like 1 Delta 1 Delta1 delta-like 1 (Drosophila) delta-like protein 1 DL1 Drosophila Delta homolog 1 H-Delta-1; Delta-like protein 1; Delta1; Drosophila Delta homolog 1; H-Delta-1
Gene Aliases: Delta; DELTA1; DL1; DLL1; UNQ146/PRO172
UniProt ID: (Human) O00548
Entrez Gene ID: (Human) 28514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.