Invitrogen
Manufacturer Code:PA519106
Catalog # PIPA519106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ATQRHLTVGEEWSQD) corresponding to the internal amino acids of DLL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: delta like-1; Delta-like protein 1; Delta1; DLL1 delta-like 1 (Drosophila) DELTA1 DL1 Delta OTTHUMP00000017690 delta (Drosophila)-like 1 delta-like 1 delta-like 1 protein; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1
Gene Aliases: Delta1; Dll1
UniProt ID: (Mouse) Q61483, (Rat) P97677
Entrez Gene ID: (Mouse) 13388, (Rat) 84010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.