Abnova
Manufacturer Code:MAB16056
Catalog # 89-998-834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PF299-1 |
Immunogen | Recombinant protein corresponding to extracellular domain of human DLK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: delta-like 1 homolog; DLK; DLK-1; FA1; Fetal antigen 1; pG2; preadipocyte factor 1; Pref-1; PREF1; Protein delta homolog 1; secredeltin; ZOG
Gene Aliases: Delta1; DLK; DLK-1; DLK1; FA1; pG2; Pref-1; PREF1; ZOG
UniProt ID: (Human) P80370
Entrez Gene ID: (Human) 8788
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.