Test
EMD Millipore
Manufacturer Code:MABN671
Catalog # MABN671MI
Quantity:
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export as a spreadsheet
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer. |
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com.
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase.
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Shared lists are a new way to save products. Create a list for each of your projects to share with collaborators like your lab manager. Then select and buy products as a team.
Creating a shared list
Click "Save to list" to create your first list, give it a name, and add the product. Access your shared lists in the Account link on Thermofisher.com. |
Collaborate
You can invite collaborators like your lab manager or procurement staff to any shared list. They can add more items, leave comments and complete the final purchase. |
Export your list as a spreadsheet for easy documentation or hand off to your lab’s designated buyer.
Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A10 |
Immunogen | Purified recombinant fragment of human DLK1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | &minus20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
Mouse Monoclonal Antibody
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.