Proteintech
Manufacturer Code:106361AP150UL
Catalog # 50-559-617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DLK1 Fusion Protein Ag0991 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipocyte differentiation inhibitor protein; delta-like 1 homolog; DLK DLK 1 DLK1 FA1 pG2 Pref 1 PREF1 Protein delta homolog 1 ZOG; DLK-1; FA1; Fetal antigen 1; pG2; Preadipocyte factor 1; preadipocyte factor-1; Pref-1; Protein delta homolog 1; secredeltin; stromal cell derived protein 1
Gene Aliases: AW742678; Delta1; DLK; DLK-1; DLK1; DlkI; FA1; Ly107; Peg9; pG2; Pref-1; PREF1; Scp-1; SCP1; ZOG
UniProt ID: (Human) P80370, (Mouse) Q09163, (Rat) O70534
Entrez Gene ID: (Human) 8788, (Mouse) 13386, (Rat) 114587
Molecular Function:
extracellular matrix protein
extracellular matrix structural protein
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.