Novus Biologicals
Manufacturer Code:NBP128739
Catalog # NBP128739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1325 and 1375 of human discs large homolog 5 using the numbering given in entry NP_004738.3 (GeneID 9231). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 5; discs large homolog 5 (Drosophila) KIAA0583large (Drosophila) homolog 5 LP-DLG; discs large protein LP-DLG; Discs large protein P-dlg; discs, large homolog 5; Disks large homolog 5; large type of P-DLG; Placenta and prostate DLG
Gene Aliases: DLG5; KIAA0583; LP-DLG; P-DLG5; PDLG
UniProt ID: (Human) Q8TDM6
Entrez Gene ID: (Human) 9231
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.