Proteintech
Manufacturer Code:156871AP150UL
Catalog # 50-173-0405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DLG5 Fusion Protein Ag8194 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 5; discs large protein LP-DLG; Discs large protein P dlg Disks large homolog 5 DLG5 KIAA0583 LP DLG P DLG5 PDLG Placenta and prostate DLG; Discs large protein P-dlg; discs, large homolog 5; Disks large homolog 5; large type of P-DLG; Placenta and prostate DLG
Gene Aliases: 4933429D20Rik; DLG5; KIAA0583; LP-DLG; mKIAA0583; P-DLG5; PDLG; T25557
UniProt ID: (Human) Q8TDM6, (Mouse) E9Q9R9
Entrez Gene ID: (Human) 9231, (Mouse) 71228
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.