OriGene
Manufacturer Code:CF809015
Catalog # 50-166-9223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI12A5 |
Immunogen | Full length human recombit protein of human DLG4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 4; Disks large homolog 4; post-synaptic density protein 95; Postsynaptic density protein 95; PSD-95; PSD95 SAP-90 SAP90; SAP-90; Synapse-associated protein 90; Tax interaction protein 15
Gene Aliases: DLG4; PSD95; SAP-90; SAP90
UniProt ID: (Human) P78352
Entrez Gene ID: (Human) 1742
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.