Novus Biologicals
Manufacturer Code:NBP288994
Catalog # NB036585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RPI 197.4 |
Immunogen | Recombinant rat DLG1(N-terminal fragment). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 1; discs large homolog 1 (Drosophila) disks large homolog 1 DKFZp761P0818 DKFZp781B0426 hDlg large (Drosophila) homolog 1 presynaptic protein SAP97; discs large homolog 1, scribble cell polarity complex component; discs, large homolog 1; discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; Disks large homolog 1; hDlg; presynaptic protein SAP97; SAP-97; Synapse-associated protein 97
Gene Aliases: dJ1061C18.1.1; DLG1; DLGH1; hdlg; SAP-97; SAP97
UniProt ID: (Human) Q12959
Entrez Gene ID: (Human) 1739
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.