Invitrogen
Manufacturer Code:PA570397
Catalog # PIPA570397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human DLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aei; after eight protein; branched chain alpha-keto acid dehydrogenase complex subunit E3; diaphorase; Dihydrolipoamide dehydrogenase; dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex); dihydrolipoamide: NAD+ oxidoreductase; dihydrolipoyl dehydrogenase mitochondrial-like protein; Dihydrolipoyl dehydrogenase, mitochondrial; DLDH E3 GCSL LAD PHE3 E3 component of pyruvate dehydrogenase complex 2-oxo-glutarate complex branched chain keto acid dehydrogenase complex diaphorase dihydrolipoyl dehydrogenase mitochondrial glycine cleavage system L protein glycine cleavage system protein L lipoamide deh; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; Glycine cleavage system L protein; glycine cleavage system protein L; lipoamide dehydrogenase; lipoamide reductase; lipoyl dehydrogenase; mitochondrial Dihydrolipoyl dehydrogenase-like protein
Gene Aliases: DLD; DLDD; DLDH; E3; GCSL; LAD; PHE3; sc:d0402
UniProt ID: (Dog) P49819, (Human) P09622, (Rat) Q6P6R2, (Mouse) O08749
Entrez Gene ID: (Dog) 403978, (Bovine) 533910, (Rabbit) 100345929, (Horse) 100050920, (Human) 1738, (Guinea pig) 100721798, (Rat) 298942, (Mouse) 13382, (Zebrafish) 30138
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.