Invitrogen
Manufacturer Code:PA576640
Catalog # PIPA576640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human DLC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP 7ARHGAP7deleted in liver cancer 1Deleted in liver cancer 1 proteinDLC 1Dlc-1DLC1FLJ21120HPHP proteinKIAA1723p122 RhoGAPRHG07&lowbarHUMANRho GTPase activating protein 7Rho GTPase-activating protein 7Rho type GTPase activating protein 7Rho-type GTPase-activating protein 7StAR related lipid transfer protein 12StAR-related lipid transfer protein 12STARD 12StARD12START domain containing protein 12START domain-containing protein 12; Deleted in liver cancer 1 protein homolog; DLC-1; p122-RhoGAP; Rho GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12
Gene Aliases: A730069N07Rik; Arhgap7; dlc-1; Dlc1; HP; STARD12
UniProt ID: (Mouse) Q9R0Z9
Entrez Gene ID: (Mouse) 50768
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.