Invitrogen
Manufacturer Code:PA569821
Catalog # PIPA569821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human DLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP7 HP STARD12 p122-RhoGAP Rho-GTPase-activating protein 7 START domain-containing protein 12 StAR-related lipid transfer (START) domain containing 12 deleted in liver cancer 1 protein deleted in liver cancer 1 variant 2 rho GTPase-activating protein 7 rho-type GTPase-activating pr; deleted in liver cancer 1; Deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; HP protein; p122-RhoGAP; rho GTPase activating protein 7; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12; unm_sa1141
Gene Aliases: A730069N07Rik; ARHGAP7; DLC-1; DLC1; HP; KIAA1723; p122-RhoGAP; RhoGAP; STARD12
UniProt ID: (Human) Q96QB1, (Bovine) A7E300, (Dog) B9VTT2, (Mouse) Q9R0Z9
Entrez Gene ID: (Rabbit) 100352546, (Human) 10395, (Bovine) 511433, (Dog) 475607, (Horse) 100052317, (Mouse) 50768, (Rat) 58834, (Zebrafish) 567965
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.