Invitrogen
Manufacturer Code:PA518290
Catalog # PIPA518290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SVAIRKRSWEEHC) corresponding to the N-terminus amino acids of DLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; DLC1 HP ARHGAP7 STARD12 FLJ21120 p122-RhoGAP deleted in liver cancer 1 Rho-GTPase-activating protein 7 START domain containing protein 12 StAR-related lipid transfer protein 12 StAR-related lipid transfer (START) domain containing 12 deleted in liver cancer 1 variant 2; HP protein; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12
Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12
UniProt ID: (Human) Q96QB1
Entrez Gene ID: (Human) 10395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.