Novus Biologicals
Manufacturer Code:NBP216173
Catalog # NBP216173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human DLC1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP7 STARD12 deleted in liver cancer 1 variant 2 deleted in liver cancer 1 Deleted in liver cancer 1 protein DLC-1 FLJ21120 HP HP protein KIAA1723 p122-RhoGAP rho GTPase-activating protein 7 Rho-GTPase-activating protein 7 Rho-type GTPase-activating protein 7 StARD12 StAR-related lipid transfer (START) domain containing 12 StAR-related lipid transfer protein 12 START domain-containing protein 12; Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; HP protein; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12
Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12
UniProt ID: (Human) Q96QB1
Entrez Gene ID: (Human) 10395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.