Abnova
Manufacturer Code:PAB6661
Catalog # 89-132-575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human DLC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of DLC1.
For Research Use Only
Protein Aliases: ARHGAP7; Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; FLJ21120; HP; HP protein; p122-RhoGAP; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; STARD12; START domain-containing protein 12
Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12
UniProt ID: (Human) Q96QB1
Entrez Gene ID: (Human) 10395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.