Proteintech
Manufacturer Code:154601AP150UL
Catalog # 50-173-0246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DLC1 Fusion Protein Ag7732 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP7 deleted in liver cancer 1 DLC 1 DLC1 HP HP protein KIAA1723 p122 RhoGAP STARD12; Deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; HP protein; p122-RhoGAP; rho GTPase activating protein 7; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12
Gene Aliases: A730069N07Rik; ARHGAP7; dlc-1; DLC1; HP; KIAA1723; p122-RhoGAP; RhoGAP; STARD12
UniProt ID: (Human) Q96QB1, (Mouse) Q9R0Z9
Entrez Gene ID: (Human) 10395, (Mouse) 50768, (Rat) 58834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.