Abnova
Manufacturer Code:H00010395K
Catalog # 89-204-599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human DLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human DLC1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ARHGAP7 FLJ21120 HP STARD12 p122-RhoGAP; Deleted in liver cancer 1 protein; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC-1; HP protein; Rho GTPase-activating protein 7; Rho-GTPase-activating protein 7; Rho-type GTPase-activating protein 7; StAR-related lipid transfer (START) domain containing 12; StAR-related lipid transfer protein 12; StARD12; START domain-containing protein 12
Gene Aliases: ARHGAP7; DLC1; HP; KIAA1723; p122-RhoGAP; STARD12
UniProt ID: (Human) Q96QB1
Entrez Gene ID: (Human) 10395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.