Invitrogen
Manufacturer Code:MA517067
Catalog # PIMA517067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8A5C6 |
Immunogen | Purified recombit fragment of human DKK3 (amino acids: 91-350 ) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dickkopf 3 homolog; dickkopf homolog 3; Dickkopf-3; Dickkopf-related protein 3; dkk-3; hDkk-3; regulated in glioma; RIG-like 5-6; RIG-like 7-1
Gene Aliases: DKK3; REIC; RIG; UNQ258/PRO295
UniProt ID: (Human) Q9UBP4
Entrez Gene ID: (Human) 27122
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.