Proteintech
Manufacturer Code:103651AP150UL
Catalog # 50-172-6247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DKK3 Fusion Protein Ag0417 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dickkopf 3 Dickkopf related protein 3 Dkk 3 DKK3 hDkk 3 REIC RIG; dickkopf 3 homolog; dickkopf homolog 3; Dickkopf-3; Dickkopf-related protein 3; dkk-3; hDkk-3; mDkk-3; regulated in glioma; RIG-like 5-6; RIG-like 7-1
Gene Aliases: AW061014; C87148; DKK3; REIC; RIG; UNQ258/PRO295
UniProt ID: (Human) Q9UBP4, (Mouse) Q9QUN9
Entrez Gene ID: (Human) 27122, (Mouse) 50781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.